Human recombinant sorbitol dehydrogenase (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-671
PRSI91-671EA 884 EUR InStock
PRSI91-671
Human recombinant sorbitol dehydrogenase (from cells)
Enzyme
Sorbitol dehydrogenase, also known as L-iditol 2-dehydrogenase and SORD, is a member of the zinc-containing alcohol dehydrogenase family. SORD exsits in a homotetramer and binds one zinc ion per subunit. SORD is expressed in kidney and epithelial cells of both benign and malignant prostate tissue. SORD can converts sorbitol to fructose and catalyses the interconversion of polyols and their corresponding ketoses, and together with aldose reductase to make up the sorbitol pathway. SORD is up-regulated by androgens and down-regulated by castration. SORD may play a role in the sperm motility by providing an energetic source for sperm.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge