Human recombinant MAG (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-674
PRSI91-674EA 654 EUR InStock
PRSI91-674
Human recombinant MAG (from cells)
Proteine und Peptide
Human Myelin-Associated Glycoprotein,also known as MAG, Siglec-4,is a cell membrane glycoprotein that is a member of the SIGLEC family of proteins.MAG contains 4 Ig-like C2-type domains and 1 Ig-like V-type domain.MAG is believed to be involved in myelination during nerve regeneration. it is a adhesion molecule in postnatal neural development that mediates sialic-acid dependent cell-cell interactions between neuronal and myelinating cells and Preferentially binds to alpha-2,3-linked sialic acid.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge