Human recombinant IL-22 binding (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-699
PRSI91-699EA 654 EUR InStock
PRSI91-699
Human recombinant IL-22 binding (from cells)
Proteine und Peptide
Interleukin-22 Receptor Subunit alpha-2 (IL22RA2) belongs to the type II cytokine receptor family. IL22RA2 is a secreted protein and contains three fibronectin type-III domains. IL22RA2 is widely expressed in many tissues. IL22RA2 functions as an IL22 antagonist and may be important in the regulation of inflammatory response. Three alternatively spliced transcript variants encoding distinct isoforms have been described.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge