Human recombinant FLRT3 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-706
PRSI91-706EA 654 EUR InStock
PRSI91-706
Human recombinant FLRT3 (from cells)
Proteine und Peptide
Leucine-Rich Repeat Transmembrane Protein FLRT3 (FLRT3) is a member of the fibronectin leucine rich transmembrane protein (FLRT) family. Proteins in this family play an role in cell adhesion and/or receptor signalling. FLRT3 is a single-pass type I membrane protein and contains one fibronectin type-III domain, ten LRR (leucine-rich) repeats, one LRRCT domain, and one LRRNT domain. FLRT3 may have a function in cell adhesion and/or receptor signaling. FLRT3 may regulate cellular adhesion between the epithelial apical ridge and the underlying mesenchyme and in establishing the dorso-ventral position of the ridge.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge