Human recombinant Apolipoprotein A-IV (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-720
PRSI91-720EA 852 EUR InStock
PRSI91-720
Human recombinant Apolipoprotein A-IV (from cells)
Proteine und Peptide
Apolipoprotein A4 (APOA4) is a secreted protein that belongs to the apolipoprotein A1/A4/E family. Apoa-IV is a major component of HDL and chylomicrons. APOA4 is secreted into circulation on the surface of newly synthesized chylomicron particles. APOA4 play a role in the regulation of appetite and satiety in rodent models. APOA4 involved in chylomicrons and VLDL secretion and catabolism and required for efficient activation of lipoprotein lipase by ApoC-II. In addition, APOA4 is a potent activator of lecithin-cholesterol acyltransferase in vitro.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge