Human recombinant Mucin 15 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-727
PRSI91-727EA 852 EUR InStock
PRSI91-727
Human recombinant Mucin 15 (from cells)
Proteine und Peptide
Mucin-15 is a single-pass type I membrane protein member of the Mucin family. Mucins are a family of high molecular weight, heavily glycosylated proteins (glycoconjugates) produced by epithelial tissues in most metazoans. A key characteristic of Mucins is their ability to form gels. Therefore they are a key component in most gel-like secretions, serving functions from lubrication to cell signalling to forming chemical barriers. Mucin-15 is expressed in many tissues. Mucin-15 is highly glycosylated (N- and O-linked carbohydrates). Mucin-15 may play a role in the cell adhesion to the extracellular matrix.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge