Human recombinant sialate O-Acetylesterase (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-729
PRSI91-729EA 884 EUR InStock
PRSI91-729
Human recombinant sialate O-Acetylesterase (from cells)
Enzyme
Sialate O-Acetylesterase (SIAE) belongs to the family of hydrolases, specifically those acting on carboxylic ester bonds. SIAE is widely expressed with high expression levels in the testis, prostate, and colon. SIAE catalyses N-acetyl-O-acetylneuraminate and H2O to N-acetylneuraminate and acetate. SIAE removes O-acetyl ester groups from position 9 of the parent sialic acid, N-acetylneuraminic acid. SIAE down-regulates B lymphocyte antigen receptor signaling (involving CD22), and is required for immunological tolerance. Loss of function mutations in SIAE are much more frequently found in humans with autoimmune diseases especially rheumatoid arthritis and type 1 diabetes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge