Human Recombinant CD160 (from Cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-744
PRSI91-744EA 409 EUR InStock
PRSI91-744
Human Recombinant CD160 (from Cells)
Proteine und Peptide
CD160 antigen is a Lipid-anchor that exists as a disulfide-linked homomultimer. CD160 contains one Ig-like V-type domain. The human CD160 precursor is a cysteine-rich, glycosylphosphatidylinositol-anchored protein of 181 amino acids with a single Ig-like domain. It is weakly homologous to KIR2DL4. CD160 is expressed in the spleen, peripheral blood, and small intestine. Its expression is tightly associated with peripheral blood NK cells and CD8 T lymphocytes with cytolytic effector activity. CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge