Human recombinant Collectin 11 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-761
PRSI91-761EA 588 EUR InStock
PRSI91-761
Human recombinant Collectin 11 (from cells)
Proteine und Peptide
Collectin-11 is a secreted protein that belongs to the COLEC10/COLEC11 family. Collectin-11 contains one C-type lectin domain and one collagen-like domain. Collectins play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. Collectin-11 binds to various sugars including fucose and mannose, but does not bind to glucose, N-acetylglucosamine and N-acetylgalactosamine. It has a higher affinity for fucose compared to mannose. Collectin-11 binds lipopolysaccharides (LPS). It also involved in fundamental development serving as a guidance cue for neural crest cell migration. Defects in Collectin-11 are the cause of 3MC syndrome type 2 (3MC2).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge