Human recombinant Lecithin-cholesterol acyltransferase (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-786
PRSI91-786EA 760 EUR InStock
PRSI91-786
Human recombinant Lecithin-cholesterol acyltransferase (from cells)
Enzyme
Lipase family. The gene encoding this protein is expressed mainly in brain, liver and testes,followed by secreting into plasma and cerebral spinal fluid. The esterification of cholesterol is required for cholesterol transport. LCAT is a central enzyme in the extracellular metabolism of plasma lipoproteins. Defects in LCAT are the cause of lecithin-cholesterol acyltransferase deficiency (LCATD) and a cause of fish-eye disease (FED).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge