Human recombinant SEMA4B (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-802
PRSI91-802EA 884 EUR InStock
PRSI91-802
Human recombinant SEMA4B (from cells)
Proteine und Peptide
The class 4 semaphorins are integral membrane proteins that are widely expressed throughout the nervous system. SEMA4B is a single-pass type I membrane protein and contains one Ig-like C2-type (immunoglobulin-like) domain, one PSI domain and one Sema domain. Human SEMA4B is expressed in neurons. SEMA4B inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. SEMA4B negatively regulates basophil-mediated Th2 and humoral memory responses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge