Human recombinant CYR61 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-813
PRSI91-813EA 588 EUR InStock
PRSI91-813
Human recombinant CYR61 (from cells)
Proteine und Peptide
Protein CYR61, also known as CCN family member 1, Cysteine-rich angiogenic inducer 61,Insulin-like growth factor-binding protein 10 , GIG1, CYR61, CCN1 and IGFBP10, belongs to the CCN family, CYR61 is a secreted protein and contains one CTCK (C-terminal cystine knot-like) domain,one IGFBP N-terminal domain,one TSP type-1 domain and one VWFC domain. CYR61 promotes cell proliferation, chemotaxis, angiogenesis and cell adhesion. CYR61 plays important roles in inflammation and tissue repair. CYR61 is associated with diseases related to chronic inflammation, including rheumatoid arthritis, atherosclerosis, diabetes-related nephropathy and retinopathy, and many different forms of cancers.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge