Human Recombinant ADAM-like decysin-1 (from Cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-838
PRSI91-838EA 588 EUR InStock
PRSI91-838
Human Recombinant ADAM-like decysin-1 (from Cells)
Proteine und Peptide
ADAM DEC1 protein is expressed highly in the small intestine and appendix, moderately in lymph node, mucosal lining of the colon, thymus, spleen and very weakly in the bone marrow. ADAM DEC1 is induced during DC maturation and up-regulated in response to T-cell signals. It may play an important role in the control of the immune response and during pregnancy.
  • Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays.
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge