Human recombinant ketohexokinase (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-843
PRSI91-843EA 654 EUR InStock
PRSI91-843
Human recombinant ketohexokinase (from cells)
Enzyme
Ketohexokinase, also known as Hepatic fructokinase, is a member of the carbohydrate kinase PfkB family. It exits as a homodimer and most abundant in liver, kidney, gut, spleen and pancreas. Low levels also found in adrenal, muscle, brain and eye.This enzyme catalyses conversion of fructose to fructose-1-phosphate. It is the first enzyme with a specialised pathway that catabolises dietary fructose. Defects in KHK are the cause of fructosuria.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge