Human recombinant MANF (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-878
PRSI91-878EA 588 EUR InStock
PRSI91-878
Human recombinant MANF (from cells)
Proteine und Peptide
Mesencephalic astrocyte-derived neurotrophic factor(MANF) is a secreted protein which belongs to the ARMET family. MANF selectively promotes the survival of dopaminergic neurons of the ventral mid-brain. It modulates GABAergic transmission to the dopaminergic neurons of the substantia nigra. MANF enhances spontaneous, as well as evoked, GABAergic inhibitory postsynaptic currents in dopaminergic neurons. MANF inhibits cell proliferation and endoplasmic reticulum (ER) stress-induced cell death. The N-terminal region of ARMET may be responsible for neurotrophic activity while the C-terminal region may play a role in the ER stress response. MANF reduces endoplasmic reticulum (ER) stress and has neurotrophic effects on dopaminergic neurons. Intracortical delivery of recombinant MANF protein protects tissue from ischemic brain injury. MANF has been described as a survival factor for dopaminergic neurons. MANF and a homologous protein, the conserved dopamine neurotrophic factor (CDNF), form a novel evolutionary conserved family of neurotrophic factors. MANF expression was widespread in the nervous system and non-neuronal tissues.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge