Human recombinant TWEAK receptor (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-900
PRSI91-900EA 588 EUR InStock
PRSI91-900
Human recombinant TWEAK receptor (from cells)
Proteine und Peptide
Tumor necrosis factor receptor superfamily member 12A(TNFRSF12A) is also known as Fibroblast growth factor-inducible immediate-early response protein 14, FN14, CD266 antigen and tweak-receptor. TNFRSF12A is a single-pass type I membrane protein, including a 27 aa signal peptide, a 53 aa extracellular domain, a 21 aa transmembrane domain and a 28 aa cytoplasmic domain. TNFRSF12A is highly expressed in heart, placenta and kidney. TNFRSF12A can be induced by FGF1 and phorbol ester. TNFRSF12A binds to TWEAK/TNFSF12A to initiate a signal transduction cascade, causing different cellular responses such as cell death, cell proliferation, and angiogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge