Human recombinant ERBB3 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-962
PRSI91-962EA 588 EUR InStock
PRSI91-962
Human recombinant ERBB3 (from cells)
Proteine und Peptide
Receptor tyrosine-protein kinase erbB-3 is an enzyme that in humans is encoded by the ERBB3 gene. This gene encodes a member of the epidermal growth factor receptor (EGFR) family of receptor tyrosine kinases. ERBB3 belongs to the protein kinase superfamily,tyr protein kinase family and EGF receptor subfamily.It contains 1 protein kinase domain and it is expressed in Epithelial tissues and brain. This membrane-bound protein has a neuregulin binding domain but not an active kinase domain. It therefore can bind this ligand but not convey the signal into the cell through protein phosphorylation. However, it does form heterodimers with other EGF receptor family members which do have kinase activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge