Human Recombinant NGAL (from Cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-967
PRSI91-967EA 588 EUR InStock
PRSI91-967
Human Recombinant NGAL (from Cells)
Proteine und Peptide
LCN2 is iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development. LCN2 binds iron through association with 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin, and delivers or removes iron from the cell, depending on the context. LCN2 is involved in apoptosis due to interleukin-3 (IL3) deprivation: iron-loaded form increases intracellular iron concentration without promoting apoptosis, while iron-free form decreases intracellular iron levels, inducing expression of the proapoptotic protein BCL2L11/BIM, resulting in apoptosis. LCN2 is involved in innate immunity, possibly by sequestrating iron, leading to limit bacterial growth.
  • Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays.
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge