Human recombinant SOD1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-985
PRSI91-985EA 884 EUR InStock
PRSI91-985
Human recombinant SOD1 (from E. coli)
Proteine und Peptide
Superoxide Dismutase [Cu-Zn] (SOD1) is a soluble cytoplasmic and mitochondrial intermembrane space protein that belongs to the Cu-Zn superoxide dismutase family. SOD1 binds copper and zinc ions and is one of three isozymes responsible for destroying free superoxide radicals in the body. SOD1 neutralizes supercharged oxygen molecules, which can damage cells if their levels are not controlled. The enzyme protects the cell against dangerous levels of superoxide. Zinc binding promotes dimerization and stabilizes the native form. Mutations in SOD1 cause a form of familial amyotrophic lateral sclerosis. Defects in SOD1 are the cause of amyotrophic lateral sclerosis type 1 (ALS1) which is a familial form of amyotrophic lateral sclerosis, a neurodegenerative disorder affecting upper and lower motor neurons and resulting in fatal paralysis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge