Human recombinant Autophagy 3 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91-995
PRSI91-995EA 343 EUR InStock
PRSI91-995
Human recombinant Autophagy 3 (from E. coli)
Proteine und Peptide
Ubiquitin-Like-Conjugating Enzyme ATG3 (ATG3) is widely expressed and has highly levels in heart, skeletal muscle, kidney, liver and placenta. ATG3 as a E2-like enzyme, involves in autophagy and mitochondrial homeostasis. ATG3 catalyzes the conjugation of ATG8-like proteins to PE which is essential for autophagy. As an autocatalytic E2-like enzyme, ATG3 also can catalyzes the conjugation of ATG12 to itself which palys a role in mitochondrial homeostasis but not in autophagy.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge