Human recombinant Aldo-Keto Reductase 1C4 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-002
PRSI92-002EA 884 EUR InStock
PRSI92-002
Human recombinant Aldo-Keto Reductase 1C4 (from E. coli)
Enzyme
Aldo-Keto Reductase 1C4/AKR1C4 is a member of the aldo/keto reductase family that consists of more than 40 known enzymes and proteins. AKR1C4 has highly expressed in Liver. It can catalyses the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. AKR1C4 catalyses the transformation of the potent androgen dihydrotestosterone (DHT) into the less active form, 5- alpha-Androstan-3- alpha,17- beta -diol (3- alpha-diol). In addition, AKR1C4 also has some 20- alpha-Hydroxysteroid Dehydrogenase activity.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge