Human recombinant 4HPPD (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-016
PRSI92-016EA 884 EUR InStock
PRSI92-016
Human recombinant 4HPPD (from E. coli)
Proteine und Peptide
4-Hydroxyphenylpyruvate Dioxygenase (4HPPD) belongs to the 4HPPD family. 4HPPD is a key enzyme in the degradation of tyrosine, which catalyzes the second reaction in the catabolism of tyrosine the conversation of 4-hydroxyphenylpyruvate to homogentisate. 4HPPD exists in homodimer forms, which uses zinc as a cofactor to catalyze the third step in the conversion of L-phenylalanine to fumarate and acetoacetic acid. When the active 4HPPD enzyme concentration is low in the human body, it results in high levels of tyrosine concentration in the blood, which can cause mild mental retardation at birth, and degradation in vision as a patient grows older.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge