Human recombinant BCAS2 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-031
PRSI92-031EA 852 EUR InStock
PRSI92-031
Human recombinant BCAS2 (from E. coli)
Proteine und Peptide
Breast Carcinoma-Amplified Sequence 2 (BCAS2) is a member of the SPF27 family. BCAS2 is a nuclear protein and widely expressed in many rtissues. BCAS2 is identified as being overexpressed in various breast cancer cell lines. BCAS2 is a component of the spliceosome, taking part in the removal of introns from mRNA precursors. BCAS2 interacts with estrogen receptor alpha and beta, thyroid hormone receptor beta, peroxisome proliferator-activated receptor gamma. BCAS2 functions as an ER co-activator and is capable of enhancing ER-mediated transcription.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge