Human recombinant MBIPP (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-032
PRSI92-032EA 852 EUR InStock
PRSI92-032
Human recombinant MBIPP (from E. coli)
Proteine und Peptide
MAP3K12-binding inhibitory protein 1 (MBIP) is a 39kD protein high expression in the heart and lung. It is a component of the ADA2A-containing complex (ATAC) complex, a complex with histone acetyltransferase activity on histones H3 and H4, and composed of CSRP2BP, KAT2A, TADA2L, TADA3L, ZZ3, MBIP, WDR5, YEATS2, CCDC101 and DR1. In the complex, it probably interacts directly with KAT2A, CSRP2BP and WDR5. It’s function to inhibit the MAP3K12 activity to induce the activation of the JNK/SAPK pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge