Human recombinant PPP1CC (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-040
PRSI92-040EA 982 EUR InStock
PRSI92-040
Human recombinant PPP1CC (from E. coli)
Proteine und Peptide
Serine/Threonine-Protein Phosphatase PP1-Υ Catalytic Subunit (PPP1CC) is a member of the PPP phosphatase family. It is essential for cell division, participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1CC colocalizes with SPZ1 in the nucleus, with URI1 at mitochondrion, Rapidly exchanges between the nucleolar, nucleoplasmic and cytoplasmic compartments. As a cofactor, PPP1CC binds one iron ion and one manganese ion per subunit.. In addition, PPP1CC may play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca2+/calmodulin dependent protein kinase II.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N, C-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge