Human recombinant CD72 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-070
PRSI92-070EA 852 EUR InStock
PRSI92-070
Human recombinant CD72 (from E. coli)
Proteine und Peptide
B-Cell Differentiation Antigen CD72 (CD72) is a single-pass type II membrane protein. CD72 exists as a disulfide-linked homodimer and contains one C-type lectin domain. CD72 is expressed on B lineage cells, NK cells, monocytes, dendritic cells, and mast cells. CD72 is a ligand for CD5. CD72 associates with CD5, interacts with PTPN6/SHP-1 and plays a role in B-cell proliferation and differentiation. CD72 associates with CD79A in the B cell antigen receptor (BCR) complex following antigen stimulation and dampens BCR signaling through interactions with the phosphatase SHP-1.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Trx, 6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge