Human recombinant ETS1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-104
PRSI92-104EA 852 EUR InStock
PRSI92-104
Human recombinant ETS1 (from E. coli)
Proteine und Peptide
ETS1 Protein (ETS1) is a nuclear protein that belongs to the ETS family. Members of this family recognize the core consensus DNA sequence GGAA/T in target genes. Proteins function either as transcriptional activators or repressors of numerous genes. They are involved in stem cell development, cell senescence and death, and tumorigenesis. ETS1 is a transcription factor, containing one ETS DNA-binding domain and one PNT (pointed) domain. it has been shown to interact with TTRAP, UBE2I and Death Associated Protein 6.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge