Human recombinant uridine phosphorylase 1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-122
PRSI92-122EA 654 EUR InStock
PRSI92-122
Human recombinant uridine phosphorylase 1 (from E. coli)
Enzyme
Uridinephosphorylase 1 (UPP1) is a member of the family of pentosyltransferase. UPP1 catalyses the reversible phosphorolysis of uridine to uracil. The expression levels and the enzymatic activity of UPP1 are higher in human solid tumors than in adjacent normal tissues. The high level of UPP1 expression in some tumors makes it a potential prognosticfactor for some cancers, such as oral squamous cell carcinoma. UPP1 is important for the homeostatic regulation of intracellular and plasma uridine concentratios. UPP1 plays an important role in the pyrimidine salvage pathway through its catalysis of the reversible phosphorolysis of uridine to uracil.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge