Human recombinant Peroxiredoxin 3 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-157
PRSI92-157EA 654 EUR InStock
PRSI92-157
Human recombinant Peroxiredoxin 3 (from E. coli)
Proteine und Peptide
Thioredoxin-Dependent Peroxide Reductase Mitochondrial (PRDX3) is an enzyme that belongs to the AhpC/TSA family. Human and mouse PRDX3 genes are highly conserved, and they map to the regions syntenic between mouse and human chromosomes. Human PRDX3 protein has an antioxidant function and is localised in the mitochondrion. PRDX3 is involved in redox regulation of the cell. PRDX3 protects radical-sensitive enzymes from oxidative damage by a radical-generating system. It acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge