Human recombinant Casein kinase I gamma-2 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-197
PRSI92-197EA 654 EUR InStock
PRSI92-197
Human recombinant Casein kinase I gamma-2 (from E. coli)
Proteine und Peptide
CSNK1G2 is a cytoplasmic protein which contains one protein kinase domain. It is amember of the CK1 Ser/Thr protein kinase family of the large casein kinase I (CKI) subfamily. CSNK1G2 participates in Wnt signalling, involves in brain development and vesicular trafficking and neurotransmitter releasing from small synaptic vesicles. It regulates fast synaptic transmission mediated by glutamate. SMAD3 phosphorylation promotes its ligand-dependent ubiquitination and subsequent proteasome degradation, thus inhibiting SMAD3-mediated TGF-beta responses.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge