Human recombinant UPRT (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-199
PRSI92-199EA 884 EUR InStock
PRSI92-199
Human recombinant UPRT (from E. coli)
Proteine und Peptide
UPRT is a cytoplasmic enzyme which belongs to the UPRTase family. UPRT is highly expressed in leukocytes, liver, spleen and thymus, with lower expression in brain, lung and skeletal muscle. UPRTcatalyses the conversion of uracil and 5-phosphoribosyl-1-R-diphosphate to uridine monophosphate (UMP). This reaction is an important part of nucleotide metabolism, specifically the pyrimidine salvage pathway. UPRT is a potential target for rational design of drugs to treat parasitic infections and cancer
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge