Human recombinant Copine-1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-213
PRSI92-213EA 620 EUR InStock
PRSI92-213
Human recombinant Copine-1 (from E. coli)
Proteine und Peptide
Copine-1(CPNE1) encodes a calcium-dependent protein which belongs to the copine family. CPNE1contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, CPNE1 does not contain a predicted signal sequence or transmembrane domains. CPNE1 may regulate molecular events at the interface of the cell membrane and cytoplasm. CPNE1 has a broad tissue distribution and it may function in membrane trafficking.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge