Human recombinant Aminoacylase-3 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-310
PRSI92-310EA 654 EUR InStock
PRSI92-310
Human recombinant Aminoacylase-3 (from E. coli)
Enzyme
Aspartoacylase 3, also known as ACY3, N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming), Acylase III, Aminoacylase-3, Aspartoacylase-2, Aspartoacylase-2, HCV core-binding protein 1 and ASPA2, is a member of the Aspartoacylase subfamily. ACY3 plays an important role in deacetylating mercapturic acids in kidney proximal tubules and acts on N-acetyl-aromatic amino acids.ACY3 is located in the cytoplasm of S2 and S3 proximal tubules and the apical domain of S1 proximal tubules. ACY3 protein is also expressed at low levels in stomach, testis, heart, brain, lung and liver, and may function as an HCV (Hepatitis C virus) core binding protein.
Fusion-Tag: N-6 His tag.
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge