Mouse Recombinant 4-1BB
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-374
PRSI92-374EA 588 EUR InStock
PRSI92-374
Mouse Recombinant 4-1BB
Proteine und Peptide
Tumor necrosis factor receptor superfamily member 9(TNFRSF9) is a member of the tumor necrosis factor (TNF) receptor family. It can be induced by lymphocyte activation (ILA) and is expressed by activated T cells, but to a larger extent on CD8 than on CD4 T cells. In addition, TNFRSF9 expression is found on dendritic cells, follicular dendritic cells, natural killer cells, granulocytes and cells of blood vessel walls at sites of inflammation. As receptor for TNFSF9/4-1BBL, it can activate T cells and the cross-linking of this protein can enhance T cell proliferation, IL-2 secretion survival and cytolytic activity. Further, it can enhance immune activity to eliminate tumors in mice.
  • Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays.
Achtung! Bitte beachten: For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge