Human recombinant SEMA4D (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-381
PRSI92-381EA 588 EUR InStock
PRSI92-381
Human recombinant SEMA4D (from cells)
Proteine und Peptide
Semaphorin-4D is also known as A8,BB18, GR3, CD100. Semaphorin-4D belongs to the semaphorin family containing 1 Ig-like C2-type domain, 1 PSI domain and 1 Sema domain. It is the cell surface receptor for PLXN1B and PLXNB2 that plays an important role in cell-cell signalling. It promotes the migration of cerebellar granule cells and of endothelial cells, regulates dendrite and axon branching and morphogenesis. Semaphorin-4D Plays a role in the immune system; Promotes signalling via SRC and PTK2B/PYK2, which then mediates activation of phosphatidylinositol 3-kinase and of the AKT1 signalling cascade.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge