Human recombinant Methylmalonyl-CoA epimerase (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-387
PRSI92-387EA 409 EUR InStock
PRSI92-387
Human recombinant Methylmalonyl-CoA epimerase (from cells)
Enzyme
Methylmalonyl-CoA epimerase, mitochondrial(MCEE)is an enzyme which belongs to the glyoxalase I family. It converts (S)-methylmalonyl-CoA to the (R) form, catalyses the following chemical reaction: (R)-methylmalonyl-CoA (S)-methylmalonyl-CoA. It plays an important role in the catabolism of fatty acids with odd-length carbon chains. This protein deficiency is an autosomal recessive inborn error of AA metabolism, involving valine, threonine, isoleucine and methionine. This organic aciduria can appear in the neonatal period with life-threatening metabolic acidosis, hyperammonemia, feeding difficulties, pancytopenia and coma.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge