Human recombinant serpin I2 (from cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-404
PRSI92-404EA 654 EUR InStock
PRSI92-404
Human recombinant serpin I2 (from cells)
Proteine und Peptide
SERPINI2 is a secreted protein of the serpin family. Serpins are a group of proteins with similar structures that were first identified as a set of proteins able to inhibit proteases. As protease inhibitors, serpins have an array of functions including regulating blood coagulation, fibrinolysis, the complement pathway, angiogenesis, inflammation, tumour suppression, extracellular matrix remodelling, and cell motility. SERPINI2 is expressed in human tissues including pancreas and adipose tissues. Mutations of human SERPINI2 can directly result in conditions such as acinar cell apoptosis and malabsorption.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge