Human recombinant sulphotransferase 1C4 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-500
PRSI92-500EA 654 EUR InStock
PRSI92-500
Human recombinant sulphotransferase 1C4 (from E. coli)
Enzyme
Human Sulfotransferase (SULT1C4) is an enzyme that in humans is encoded by the SULT1C4 gene, belongs to the sulfotransferase 1 family. SULT1C4 is expressed at high levels in fetal lung and kidney and at low levels in fetal heart, adult kidney, ovary and spinal chord. Sulfotransferase utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyse the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF). SULT1C4 plays an important role incatalyzing the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge