Mouse recombinant IL1RA (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-517
PRSI92-517EA 852 EUR InStock
PRSI92-517
Mouse recombinant IL1RA (from E. coli)
Proteine und Peptide
Interleukin-1 receptor antagonist protein (Il1rn), also known as IL-1ra, IRAP or IL1 inhibitor, is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1 alpha (IL1A) and interleukin 1 beta (IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses. The mouse Il1rn gene encodes a 178 amino acids (aa) protein with a 26 aa signal peptide. Mouse Il1rn protein shares 26% and 19% identity with its homologues IL-1 beta and IL-1 alpha, respectively. Il1rn can Inhibits the activity of interleukin-1 by binding to receptor IL1R1 and preventing its association with the coreceptor IL1RAP for signalling, but has no interleukin-1 like activity. Recently, an recombinant human Il1rn protein is used in the treatment of rheumatoid arthritis, an autoimmune disease in which IL-1 plays a key role.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge