Mouse recombinant IL-2 R alpha
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-528
PRSI92-528EA 528 EUR InStock
PRSI92-528
Mouse recombinant IL-2 R alpha
Proteine und Peptide
The interleukin 2 receptor alpha (IL2RA), also called CD25, is a type I transmembrane protein that presents on activated T cells, activated B cells, some thymocytes, myeloid precursors, and oligodendrocytes. IL2RA is a member of cytokine receptors family that utilises the common gamma chain subunit ( gamma c). IL2RA is expressed in most B-cell neoplasms, some acute nonlymphocytic leukemias, neuroblastomas, and tumour infiltrating lymphocytes. IL2RA associates with IL2RB (CD122) to form a heterodimer that can act as a high-affinity receptor for IL-2.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge