Mouse recombinant DDR2
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-640
PRSI92-640EA 461 EUR InStock
PRSI92-640
Mouse recombinant DDR2
Proteine und Peptide
Discoidin domain receptor-2 (DDR2) is a cell surface tyrosine kinase receptor that can be activated by soluble collagen and has been implicated in diverse physiological functions including organism growth and wound repair. DDR2 binds to and is activated by collagen I, II, III, V, and X, with the notable exception of basement membrane collagen IV. DDR2 is expressed in connective tissues arising from embryonic mesoderm. DDR2 regulates cell proliferation, cell adhesion, migration, and extracellular matrix remodelling.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge