Mouse recombinant MBL2
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-681
PRSI92-681EA 588 EUR InStock
PRSI92-681
Mouse recombinant MBL2
Proteine und Peptide
Mannose-binding Lectin (MBL) is an acute phase protein bearing to the family of collectins produced by the liver as a monomer that forms a triple helix. Once released in serum, it further polymerises forming dimers to octamers. The degree of serum polymerisation is critical for the biological activity of MBL. MBL has higher affinity to microbial polysaccharides or their glycoconjugates. MBL was shown earlier to bind cell surfaces of bacteria, fungi, protozoa and viruses and acts as an acute-phase plasma protein (APP) during infection and inflammation. MBL activates the lectin-complement pathway, promotes opsonophagocytosis and modulates inflammation.
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge