Mouse recombinant TREM2B
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-683
PRSI92-683EA 588 EUR InStock
PRSI92-683
Mouse recombinant TREM2B
Proteine und Peptide
Triggering receptor expressed on myeloid cells-2 (TREM-2) is a cell surface receptor primarily expressed on macrophages, osteoclasts, microglia and dendritic cells. TREM-2 is one member of the TREM family, inhibiting the releasing of inflammatory mediators, so it is an important in vivo anti-inflammatory receptor. TREM-2 consists of an 18 aa signal sequence, a 153 aa extracellular domain (ECD) with one V-type Ig-like domain, a 21 aa transmembrane (TM) domain, and a 35 aa cytoplasmic tail. A soluble form of TREM-2 (TREM-2b) created by alternate splicing diverges at aa 161. TREM-2 transduces intracellular signals through the adaptor DAP12. After binding of TREM-2 with ligand, the TREM-2/DAP12 (dead-cell-activated-receptor-associated protein)-mediated signal transduction pathway causes a series of intracellular protein tyrosine phosphorylation reactions and enzymatic reactions, which then activate the myeloid cells and participate T cell responses.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge