Human recombinant PBEF (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92-711
PRSI92-711EA 588 EUR InStock
PRSI92-711
Human recombinant PBEF (from E. coli)
Proteine und Peptide
Pre-B cell colony enhancing factor (PBEF) was originally identified as a cytokine that potentiated the clonal expansion and differentiation of pre-B cells, but it is also acknowledged to be the ubiquitous intracellular enzyme nicotinamide phosphoribosyltranferase (NAMPT) and the adipokine “visfatin”. PBEF is constitutively expressed in the fetal membranes where its greatest expression is in the amnion. It has intracellular and extracellular forms. Most of the intracellular functions of PBEF are due to its role as a Nampt which can induce angiogenesis through upregulation of VEGF and VEGFR and secretion of MCP-1. Extracellular PBEF has been shown to increase inflammatory cytokines, such as TNF- alpha, IL-1 beta , IL-16, and TGF- beta 1. PBEF also increases the production of IL-6, TNF- alpha, and IL-1 beta in CD14+ monocyctes, macrophages, and dendritic cells, enhances the effectiveness of T cells.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge