Human recombinant CD401 (from HEK293 cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96-086
PRSI96-086EA 734 EUR InStock
PRSI96-086
Human recombinant CD401 (from HEK293 cells)
Proteine und Peptide
Tumor necrosis factor receptor superfamily member 5 (TNFRSF5) is also known as CD40, is a member of the TNF receptor superfamily. The expression of CD40 is diverse. TNFRSF5 has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. CD40 is the receptor for TNFSF5/CD40LG. Defects in CD40 are the cause of immunodeficiency with hyper-IgM type 3 (HIGM3).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge