Human Recombinant Glutaredoxin 1 (from E. coli)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96-355
PRSI96-355EA 461 EUR InStock
PRSI96-355
Human Recombinant Glutaredoxin 1 (from E. coli)
Proteine und Peptide
Glutaredoxin-1 (GRX1) is also known as Thioltransferase-1 (TTase-1), GLRX, GRX, GLRX1, which belongs to the glutaredoxin family. GRX1 / GLRX contains one glutaredoxin domain, which exists in either a reduced or an oxidized form. GRX1 / GLRX has a glutathione-disulfide oxidoreductase activity in the presence of NADPH and glutathione reductase and functions as electron carriers in the glutathione-dependent synthesis of deoxyribonucleotides by the enzymeribonucleotide reductase.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge