Recombinant SCF (from HEK293 cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96-676
PRSI96-676EA 509 EUR InStock
PRSI96-676
Recombinant SCF (from HEK293 cells)
Proteine und Peptide
Kit ligand (KITLG) is also known as stem cell factor (SCF), mast cell growth factor (MGF), steel factor (SF), which belongs to the SCF family, and is a widely expressed 28 - 40 kDa type I transmembrane glycoprotein. KITLG is the ligand for the receptor-type protein-tyrosine kinase KIT. SCF / MGF plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. KITLG / SCF binding can activate several signaling pathways. KITLG / SF Promotes phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, and subsequent activation of the kinase AKT1. KITLG / SCF and KIT also transmit signals via GRB2 and activation of RAS, RAF1 and the MAP kinases MAPK1/ERK2 and/or MAPK3/ERK1. KITLG / SCF and KIT promote activation of STAT family members STAT1, STAT3 and STAT5. KITLG / SCF and KIT promote activation of PLCG1, leading to the production of the cellular signaling molecules diacylglycerol and inositol 1, 4, 5 - trisphosphate. KITLG / SCF acts synergistically with other cytokines, probably interleukins.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge