Human recombinant serpin a3 (from HEK293 cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96-684
PRSI96-684EA 686 EUR InStock
PRSI96-684
Human recombinant serpin a3 (from HEK293 cells)
Proteine und Peptide
Serpin A3 is also known as Alpha-1-antichymotrypsin (ACT or AACT), Cell growth-inhibiting gene 24/25 protein, which belongs to the serpin family. Serpin A3 can bind DNA. Although its physiological function is unclear, it can inhibit neutrophil cathepsin G and mast cell chymase, both of which can convert angiotensin-1 to the active angiotensin-2.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge