Human recombinant vasorin (from HEK293 cells)
Lieferant: PROSCI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96-765
PRSI96-765EA 686 EUR InStock
PRSI96-765
Human recombinant vasorin (from HEK293 cells)
Proteine und Peptide
Vasorin (VASN),a single-pass type I membrane protein, is also known as protein slit-like 2 and SLITL2, which contains one EGF-like domain, ten LRR (leucine-rich) repeats, one LRRCT domain and one LRRNT domain. VASN is expressed at highest levels in aorta, at intermediate levels in kidney and placenta and at lowest levels in brain, heart, liver, lung and skeletal muscle. VASN can interact selectively and non-covalently with TGF-beta, transform growth factor beta, a multifunctional peptide that controls proliferation, differentiation and other functions in many cell types. In general, VASN may act as an inhibitor of TGF-beta signaling.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge