NanoDrop One
NanoDrop One
NanoDrop One/Onec UV-Vis Spectrophotometer
Quantify and qualify DNA, RNA, and protein samples with only 1-2 µL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ One Microvolume UV-Vis spectrophotometer.
  • Prevent costly delays — identify contaminants, obtain corrected concentrations, and enjoy success in your downstream applications
  • Collect data fast — Auto-Blank and Auto-Measure modes save time while organizing data
  • Save bench space — user-friendly local control with sleek, standalone design means no PC required
Product Details